Learn More
Bioworld HUMAN GRANULOCYTE-COLONY ST
Supplier:  Bioworld 220600842
Encompass_Preferred
HUMAN GRANULOCYTE-COLONY STIMULATING FACTOR (RECOMBINANT): Recombinant Human Granulocyte-Colony Stimulating Factor (G-CSF) Source: CHO cells Background : A glycoprotein of MW 20 kDa containing internal disulfide bonds. It induces the...
Catalog No. 50-253-184