Learn More
Creative Biomart Pegfilgrastim, PEGylated Human granulocyte colony stimulating factor (GCSF)
Supplier:  Creative Biomart THP0174
Encompass_Preferred
The product is a modified form of filgrastim which has been PEGylated at the N terminus. It is a form of human G-CSF (Granulocyte colony stimulating factor) consisting of 175 residues and is produced from E. coli via bacterial fermentation.
Catalog No. 50-208-4225