Learn More
STEMCELL Technologies RosetteSep™ Human Granulocyte Depletion Cocktail, For processing: 200 mL whole blood processed
SDP
Catalog No. NC1686753
Encompass
The RosetteSep™ Human Granulocyte Depletion Cocktail is designed to deplete granulocytes from whole blood. Unwanted cells are targeted for removal with Tetrameric Antibody Complexes recognizing CD66b and glycophorin A on red blood cells (RBCs). When centrifuged over a buoyant density medium such as Lymphoprep™ (Catalog #07801), the unwanted cells pellet along with the RBCs. The granulocyte-depleted fraction is present as a highly enriched population at the interface between the plasma and the buoyant density medium.
Catalog No. NC1686753 Supplier STEMCELL Technologies Supplier No. 15664
The RosetteSep™ Human Granulocyte Depletion Cocktail is designed to deplete granulocytes from whole blood. Unwanted cells are targeted for removal with Tetrameric Antibody Complexes recognizing CD66b and glycophorin A on red blood cells (RBCs). When centrifuged over a buoyant density medium such as Lymphoprep™ (Catalog #07801), the unwanted cells pellet along with the RBCs. The granulocyte-depleted fraction is present as a highly enriched population at the interface between the plasma and the buoyant density medium.