Learn More
Abcam Human Granulysin ELISA Kit, 1 x 96 Tests
Catalog No. 502656895
Encompass_Preferred
Human Granulysin ELISA Kit is a single-wash 90-min Simplestep used to quantify Human Granulysin with a sensitivity of 8.22 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric Sandwich ELISA - 450 nm readout : works on any standard plate reader - Design your own immunoassay: we also offer the conjugation-ready antibody pair
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-265-6895 Supplier Abcam Supplier No. AB2564021X96TESTS
Add to Cart
Add to Cart
Human Granulysin ELISA Kit is a single-wash 90-min Simplestep used to quantify Human Granulysin with a sensitivity of 8.22 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric Sandwich ELISA - 450 nm readout : works on any standard plate reader - Design your own immunoassay: we also offer the conjugation-ready antibody pair
The product is subject to the following: Abcam Restricted Use Statement