Learn More
Biomatik Corporation Human Granzyme A (GZMA) High-sensitive ELISA Kit
Supplier:  Biomatik Corporation EKU10929
Encompass_Preferred
Human Granzyme A (GZMA) High-sensitive ELISA Kit for detecting GZMA in serum, plasma, tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 15.6-1,000pg/ml. Sensitivity: 5.7pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P12544. Gene ID: 3001. Target Synonyms: CTLA3; HFSP; HF; H factor; Granzyme 1; Fragmentin-1; Hanukkah factor; Cytotoxic T-Lymphocyte-Associated Serine Esterase 3; CTL tryptase; Cytotoxic T-lymphocyte proteinase 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7757