Learn More
TRIBIOSCIENCE INC Fast Human Granzyme B ELISA, 1 x 96-well plate
SDP
Catalog No. NC3454584
Encompass_Preferred
Tribioscience’s Human Granzyme B Fast ELISA is designed to quantitatively detect human Granzyme B levels in CSF, brain tissue, serum, plasma, and other biological samples. The kit uses proprietary methods to combine samples and detections in one step, rather than complex traditional methods. The detection range is from 8 to 2000 pg/mL.
Catalog No. NC3454584 Supplier TRIBIOSCIENCE INC Supplier No. TBS3208
Tribioscience’s Human Granzyme B Fast ELISA is designed to quantitatively detect human Granzyme B levels in CSF, brain tissue, serum, plasma, and other biological samples. The kit uses proprietary methods to combine samples and detections in one step, rather than complex traditional methods. The detection range is from 8 to 2000 pg/mL.