Learn More
Sino Biological Recombinant Human Granzyme B Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 10345H08H50
Encompass_Preferred
A DNA sequence encoding the proform of human Granzyme B (NP_004122.1) (Met 1-Tyr 247) was expressed with a C-terminal polyhistidine tag.
Catalog No. 50-224-6598