Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Human Granzyme B Quantikine ELISA Kit
SDP

Catalog No. DGZB00
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
DGZB00 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. DGZB00 Supplier R&D Systems™ Supplier No. DGZB00

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Human Granzyme B Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The Human Granzyme B Quantikine ELISA Kit quantifies human Granzyme B in cell culture supernates (50 μL).

Specifications

Assay Range 31.30 to 2000 pg/mL
Assay Sensitivity 4.8 pg/mL
Sample Type Cell Culture Supernatant
Specificity Natural and Recombinant Human Granzyme B
For Use With (Application) ELISA
Gene ID (Entrez) 3002
Host Species Human
Quantity 1 Kit
Sample Volume Cell Culture Supernates (50 μL)
Storage Requirements Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Synonym C11, CCPI, CGL1, CGL-1, CSPB, CSP-B, CTLA1, CTLA-1, CTSGL1, Fragmentin-2, granzyme B, Granzyme-2, GranzymeB, GRB, GrzB, GZMB, HLP, SECT
Target Granzyme B
Total Assay Time 3.5 hr.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.