Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human Granzyme H ELISA Kit

Catalog No. EH231RB
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 x 96 Tests
96 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
EH231RB 96 Tests
EH231RBX5 5 x 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. EH231RB Supplier Invitrogen™ Supplier No. EH231RB
Add to Cart
Edge
Add to Cart

ELISA

Human Granzyme H quantitates human Granzyme H in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human Granzyme H.

This gene encodes a member of the peptidase S1 family of serine proteases. Alternative splicing results in multiple transcript variants, at least one of which encodes a preproprotein that is proteolytically processed to generate a chymotrypsin-like protease. This protein is reported to be constitutively expressed in the NK (natural killer) cells of the immune system and may play a role in the cytotoxic arm of the innate immune response by inducing target cell death and by directly cleaving substrates in pathogen-infected cells. This gene is present in a gene cluster with another member of the granzyme subfamily on chromosome 14.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P20718
Assay Range 12.5-3000 pg/mL
Assay Sensitivity 12.5 pg/mL
Conjugate HRP
Immunoassay Kit Format Sandwich ELISA Kit
Product Type ELISA
Sample Type Plasma, Serum, Supernatant
Target Species Human
For Use With (Equipment) Absorbance Microplate Reader
Gene Alias Cathepsin G-like 2; cathepsin G-like 2, protein h-CCPX; CCP-X; CGL2; CGL-2; CSP-C; CTLA1; CTSGL2; Cytotoxic serine protease C; Cytotoxic T-lymphocyte proteinase; cytotoxic T-lymphocyte-associated serine esterase 1; cytotoxin serine protease-C; granzyme H; granzyme H (cathepsin G-like 2, protein h-CCPX); GZMHCathepsin G-like 2; cathepsin G-like 2, protein h-CCPX; CCP-X; CGL2; CGL-2; CSP-C; CTLA1; CTSGL2; Cytotoxic serine protease C; Cytotoxic T-lymphocyte proteinase; cytotoxic T-lymphocyte-associated serine esterase 1; cytotoxin serine protease-C; granzyme H; granzyme H (cathepsin G-like 2, protein h-CCPX); GZMH;
Gene ID (Entrez) 2999
Gene Symbol GZMH
Interassay CV <12%
Intraassay CV <10%
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye Biotin
Quantity 96 Tests
Regulatory Status RUO
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 50 μL
Shipping Condition Wet or Dry Ice
Storage Requirements Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at -20°C
Target Granzyme H
Test Time 1 hr 20 min
Total Assay Time 4 hr 45 min
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.