Learn More
Sino Biological Anti-Progranulin Antibody, Mouse Monoclonal
Supplier:  Sino Biological 10826MM1020
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human Progranulin / Granulin (rh Progranulin / Granulin; Catalog#10826-H08H; NP_002078.1; Met1-Leu593). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-241-0388