Learn More
Sino Biological Anti-Progranulin Antibody, Mouse Monoclonal
Supplier:  Sino Biological 10826MM0220
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human Granulin / GRN (rhGRN; Catalog#10826-H08H; Met 1-Leu 593; NP_002078.1). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-240-8732