Learn More
FUJIFILM BIOSCIENCES INC Human GRO-beta (CXCL2) , Recombinant Protein, Shenandoah Biotechnology, 100 ug vial
Supplier:  FUJIFILM BIOSCIENCES INC 100123100UG
Encompass_Preferred
Growth regulated protein beta (GRO-B), also known as CXCL2, is a chemokine that is secreted by macrophages, neutrophils, and monocytes at sites of inflammation. GRO-B functions as a chemoattractant for leukocytes and hematopoietic stem cells. GRO-B activity is mediated through binding the G-protein-coupled chemokine receptor CXCR2.
Catalog No. 50-900-483