Learn More
FUJIFILM BIOSCIENCES INC Human GRO-beta (CXCL2) , Recombinant Protein, Shenandoah Biotechnology, 1 mg vial
Supplier:  FUJIFILM BIOSCIENCES INC 1001231MG
Encompass_Preferred
Growth regulated protein beta (GRO-B), also known as CXCL2, is a chemokine that is secreted by macrophages, neutrophils, and monocytes at sites of inflammation. GRO-B functions as a chemoattractant for leukocytes and hematopoietic stem cells. GRO-B activity is mediated through binding the G-protein-coupled chemokine receptor CXCR2.
Catalog No. 50-900-484