Learn More
American Research Products Inc Human Growth differentiation f
Catalog No. 5000326837
Encompass
Human Growth/differentiation factor 11 (GDF11) ELISA kit from CUSABIO Detection Range 28 pg/mL-1800 pg/mL Sensitivity 7 pg/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-26837 Supplier American Research Products Inc Supplier No. CSBEL009344HU
Human Growth/differentiation factor 11 (GDF11) ELISA kit from CUSABIO Detection Range 28 pg/mL-1800 pg/mL Sensitivity 7 pg/mL Sample serum plasma tissue homogenates Method Sandwich