Learn More
Biomatik Corporation Human Growth Differentiation Factor 5 (GDF5) Instant ELISA Kit
Supplier:  Biomatik Corporation EKU11984
Encompass_Preferred
Human Growth Differentiation Factor 5 (GDF5) Instant ELISA Kit for detecting GDF5 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 15.6-1,000pg/ml. Sensitivity: 6.2pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 1h, 10min. Accession Number: P43026. Gene ID: 8200. Target Synonyms: CDMP1; LAP4; SYNS2; BMP14; Radotermin; Cartilage-Derived Morphogenetic Protein-1; Bone morphogenetic protein 14; Lipopolysaccharide-associated protein 4. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7466