Learn More
Biomatik Corporation Human Growth Differentiation Factor 15 (GDF15) CLIA Kit
Supplier:  Biomatik Corporation EKU09886
Encompass_Preferred
Human Growth Differentiation Factor 15 (GDF15) CLIA Kit for detecting GDF15 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 6.86-5,000pg/ml. Sensitivity: 2.85pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: Q99988. Gene ID: 9518. Target Synonyms: PDF; MIC1; NAG-1; PLAB; PTGFB; Macrophage inhibitory cytokine 1; NSAID-activated gene 1 protein; Placental bone morphogenetic protein; Prostate differentiation factor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7516