Learn More
American Research Products Inc Human growth differentiation f
Catalog No. 5000329770
Encompass
Human growth differentiation factor 2 GDF2 ELISA Kit from CUSABIO Detection Range 3 9 pg/mL-250 pg/mL Sensitivity 0 98 pg/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-29770 Supplier American Research Products Inc Supplier No. CSBE11769H
Human growth differentiation factor 2 GDF2 ELISA Kit from CUSABIO Detection Range 3 9 pg/mL-250 pg/mL Sensitivity 0 98 pg/mL Sample serum plasma tissue homogenates Method Sandwich