Learn More
Raybiotech Inc C-Series Human Growth Factor Antibody Array 1 Kit.
Supplier:  Raybiotech Inc AAHGF12
Encompass_Preferred
Detects 41 Human Growth Factors. Suitable for all liquid sample types. Semi-Quantitative, Sandwich-based, Membrane Antibody Array. Screen and compare protein expression levels with high specificity and sensitivity (pg/ml levels). No dedicated equipment required. Compatible with any chemiluminescent imaging system. Proteins detected: Amphiregulin, bFGF, beta-NGF, EGF, EGFR, FGF-4, FGF-6, FGF-7 (KGF), GCSF, GDNF, GM-CSF, HB-EGF, HGF, IGFBP-1, IGFBP-2, IGFBP-3, IGFBP-4, IGFBP-6, IGF-1, IGF-1 R, IGF-2, M-CSF, M-CSF R, NT-3, NT-4, PDGF R alpha, PDGF R beta, PDGF-AA, PDGF-AB, PDGF-BB, PLGF, SCF, SCF R (CD117/c-kit), TGF alpha, TGF beta 1, TGF beta 2, TGF beta 3, VEGF-A, VEGFR2, VEGFR3, VEGF-D
Catalog No. 50-194-8583