Learn More
Raybiotech Inc G-Series Human Growth Factor Array 1 Kit.
Supplier:  Raybiotech Inc GSHGF12
Encompass_Preferred
Semi-quantitative measurement of 40 human cytokines. Suitable for all liquid sample types. Semi-Quantitative, Sandwich-based, Glass Slide Antibody Array Screen and compare protein expression levels with high specificity and sensitivity (pg/ml levels). The glass slide array allows for high throughput and low sample consumption; as little as 10 µl of original sample required per array. Proteins detected: Amphiregulin, BDNF, bFGF, BMP-4, BMP-5, BMP-7, beta-NGF, EGF, EGFR, EG-VEGF (PK1), FGF-4, FGF-7 (KGF), GDF-15, GDNF, Growth Hormone, HB-EGF, HGF, IGFBP-1, IGFBP-2, IGFBP-3, IGFBP-4, IGFBP-6, IGF-1, Insulin, M-CSF R, NGFR (TNFRSF16), NT-3, NT-4, Osteoprotegerin (TNFRSF11B), PDGF-AA, PLGF, SCF, SCF R (CD117/c-kit), TGF alpha, TGF beta 1, TGF beta 3, VEGF-A, VEGFR2, VEGFR3, VEGF-D
Catalog No. 50-195-0399