Learn More
Biomatik Corporation Human Growth Factor Receptor Bound Protein 10 (Grb10) ELISA Kit
Supplier:  Biomatik Corporation EKU04602
Encompass_Preferred
Human Growth Factor Receptor Bound Protein 10 (Grb10) ELISA Kit for detecting Grb10 in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.102ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q13322. Gene ID: 2887. Target Synonyms: GRB-IR; IRBP; MEG1; RSS; GRB10 adapter protein; Insulin receptor-binding protein Grb-IR. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5138