Learn More
Creative Biomart Recombinant E. coli Shiga Like Toxin-2 Subunit B Antigen, His-tagged
Catalog No. 502084842
Encompass_Preferred
Recombinant Shiga Like Toxin-2 Subunit B from E. coli O157:H7 produced in E. coli having an Mw of 22 kDa.The Shiga Like Toxin 2 protein is fused to a 6 His tag at its N-terminus and purified by proprietary chromatographic technique.
Catalog No. 50-208-4842 Supplier Creative Biomart Supplier No. THP0329
Recombinant Shiga Like Toxin-2 Subunit B from E. coli O157:H7 produced in E. coli having an Mw of 22 kDa.The Shiga Like Toxin 2 protein is fused to a 6 His tag at its N-terminus and purified by proprietary chromatographic technique.