Learn More
Biomatik Corporation Human Growth Hormone (GH) ELISA Kit
Supplier:  Biomatik Corporation EKU10117
Encompass_Preferred
Human Growth Hormone (GH) ELISA Kit for detecting GH in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 123.5-10,000pg/ml. Sensitivity: 44.7pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Accession Number: P01241. Gene ID: 2688. Target Synonyms: GH1; GH-N; GHN; hGH-N; Somatotropin; Hygetropin; Jintropin; Kigtropin; Pituitary Growth Hormone; Growth Hormone, Normal. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-4806