Learn More
ACROBiosystems ACROBiosystems Human Growth Hormone R (GHR) Protein, His Tag
Catalog No. 502106649
Encompass_Preferred
Recombinant Protein;50UG;Human GHR, His Tag (GHR-H5222) is expressed from human 293 cells (HEK293). It contains AA Ala 27 - Tyr 264 (Accession # P10912-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 29.6 kDa. The protein migrates as 40-55 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human GH, Tag Free at 2 μg/mL (100 μL/well) can bind Human GHR, His Tag (Cat. No. GHR-H5222) with a linear range of 1-8 ng/mL (QC tested).
Catalog No. 50-210-6649 Supplier ACROBiosystems Supplier No. GHRH522250UG
Recombinant Protein;50UG;Human GHR, His Tag (GHR-H5222) is expressed from human 293 cells (HEK293). It contains AA Ala 27 - Tyr 264 (Accession # P10912-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 29.6 kDa. The protein migrates as 40-55 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human GH, Tag Free at 2 μg/mL (100 μL/well) can bind Human GHR, His Tag (Cat. No. GHR-H5222) with a linear range of 1-8 ng/mL (QC tested).