Learn More
Bioworld HUMAN GROWTH HORMONE RELEAS
Catalog No. 50253440
Encompass_Preferred
Human Growth Hormone Releasing Hormone (rHuGHRH) Source: E.coli Description : Synthetic Human GHRH is a single, non-glycosylated, polypeptide chain containing 29 amino acids and having a molecular mass of 3358 Dalton. Corresponds to ...
Catalog No. 50-253-440 Supplier Bioworld Supplier No. 220603431
Human Growth Hormone Releasing Hormone (rHuGHRH) Source: E.coli Description : Synthetic Human GHRH is a single, non-glycosylated, polypeptide chain containing 29 amino acids and having a molecular mass of 3358 Dalton. Corresponds to ...