Learn More
Raybiotech Inc Human/Mouse/Rat GRP EIA Kit.
Supplier:  Raybiotech Inc EIAGRP1
Encompass_Preferred
Optimized for serum, plasma, and cell culture supernatants. Competition-based ELISA on a 96-well strip plate.
Catalog No. 50-194-8819