Learn More
Abcam Human GRP78 BiP ELISA Kit, 96 Tests
Catalog No. 502654106
Encompass_Preferred
Human GRP78 BiP ELISA Kit is a single-wash 90-min Simplestep used to quantify Human GRP78 BiP with a sensitivity of 0.051 ng/mL. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric Sandwich ELISA - 450 nm readout : works on any standard plate reader - Design your own immunoassay: we also offer the conjugation-ready antibody pair
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-265-4106 Supplier Abcam Supplier No. AB30276596TESTS
Human GRP78 BiP ELISA Kit is a single-wash 90-min Simplestep used to quantify Human GRP78 BiP with a sensitivity of 0.051 ng/mL. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric Sandwich ELISA - 450 nm readout : works on any standard plate reader - Design your own immunoassay: we also offer the conjugation-ready antibody pair
The product is subject to the following: Abcam Restricted Use Statement