Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human GSTM1 ELISA Kit

Catalog No. EH233RBX5
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 x 96 Tests
5 x 96 Tests
96 Tests
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
EH233RBX5 5 x 96 Tests
EH233RBX10 10 x 96 Tests
EH233RB 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. EH233RBX5 Supplier Invitrogen™ Supplier No. EH233RBX5
Get Up to $3,000 in Extra Life Sciences Consumables with Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

ELISA

Human GSTM1 quantitates human GSTM1 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human GSTM1.

Cytosolic and membrane-bound forms of glutathione S-transferase are encoded by two distinct supergene families. At present, eight distinct classes of the soluble cytoplasmic mammalian glutathione S-transferases have been identified: alpha, kappa, mu, omega, pi, sigma, theta and zeta. This gene encodes a glutathione S-transferase that belongs to the mu class. The mu class of enzymes functions in the detoxification of electrophilic compounds, including carcinogens, therapeutic drugs, environmental toxins and products of oxidative stress, by conjugation with glutathione. The genes encoding the mu class of enzymes are organized in a gene cluster on chromosome 1p13. 3 and are known to be highly polymorphic. These genetic variations can change an individual's susceptibility to carcinogens and toxins as well as affect the toxicity and efficacy of certain drugs. Null mutations of this class mu gene have been linked with an increase in a number of cancers, likely due to an increased susceptibility to environmental toxins and carcinogens. Multiple protein isoforms are encoded by transcript variants of this gene.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P09488
Assay Range 2-500 ng/mL
Assay Sensitivity 2 ng/mL
Conjugate HRP
Immunoassay Kit Format Sandwich ELISA Kit
Product Type ELISA
Sample Type Plasma, Serum, Supernatant
Target Species Human
For Use With (Equipment) Absorbance Microplate Reader
Gene Alias glutathione S-alkyltransferase; glutathione S-aralkyltransferase; glutathione S-aryltransferase; glutathione S-transferase GT8.7; glutathione S-transferase M1; glutathione S-transferase Mu 1; Glutathione S-transferase Yb-1; glutathione S-transferase Yb-1 subunit; glutathione S-transferase, mu 1; glutathione-S-transferase mu type 1 (Yb1); glutathione-S-transferase, mu 1; glutathione-S-transferase, mu type 1 (Yb1); GST 1-1; GST 3-3; GST class-mu 1; GST HB subunit 4; GST Yb1; GST1; GSTA3; Gstb1; Gstb-1; GSTM1; GSTM1-1; GSTM1a-1a; GSTM1b-1b; GTH4; GTM1; H-B; HB subunit 4; MU; MU-1; pmGT10; S-(hydroxyalkyl)glutathione lyaseglutathione S-alkyltransferase; glutathione S-aralkyltransferase; glutathione S-aryltransferase; glutathione S-transferase GT8.7; glutathione S-transferase M1; glutathione S-transferase Mu 1; Glutathione S-transferase Yb-1; glutathione S-transferase Yb-1 subunit; glutathione S-transferase, mu 1; glutathione-S-transferase mu type 1 (Yb1); glutathione-S-transferase, mu 1; glutathione-S-transferase, mu type 1 (Yb1); GST 1-1; GST 3-3; GST class-mu 1; GST HB subunit 4; GST Yb1; GST1; GSTA3; Gstb1; Gstb-1; GSTM1; GSTM1-1; GSTM1a-1a; GSTM1b-1b; GTH4; GTM1; H-B; HB subunit 4; MU; MU-1; pmGT10; S-(hydroxyalkyl)glutathione lyase;
Gene ID (Entrez) 2944
Gene Symbol GSTM1
Interassay CV <12%
Intraassay CV <10%
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye Biotin
Quantity 5 x 96 Tests
Regulatory Status RUO
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 50 μL
Shipping Condition Wet or Dry Ice
Storage Requirements Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at -20°C
Target GSTM1
Test Time 1 hr 20 min
Total Assay Time 4 hr 45 min
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.