Learn More
Sino Biological Recombinant Anti-GSTM2 Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 12042R011100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human GSTM2 (rh GSTM2; Catalog#12042-H07E; P28161; Met1-Lys218).
Catalog No. 50-240-8103