Learn More
Sino Biological Anti-GSTM2 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 12042RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human GSTM2 (rh GSTM2; Catalog#12042-H07E; P28161; Met1-Lys218). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-240-8611