Learn More
Cusabio Technology LLC HUMAN GSTO1 ELISA KIT
Catalog No. 5000049105
Encompass_Preferred
This kit detects GSTO1 in human serum plasma tissue homogenates and cell lysates using a sensitive sandwich ELISA method It offers a broad detection range of 31 2 pg/mL to 2000 pg/mL with a high sensitivity of 7 8 pg/mL ensuring accurate quantification of low-abundance targets The assay requires a short incubation period of 1 to 5 hours and a modest sample volume of 50-100 L per test Detection is performed at 450 nm and the kit is designed for reliable performance in both research and clinical settings Its standardized protocol supports consistent results across diverse sample types making it suitable for studies focused on oxidative stress detoxification pathways or biomarker analysis related to GSTO1 expression
Catalog No. 50-000-49105 Supplier Cusabio Technology LLC Supplier No. CSBEL009986HU
This kit detects GSTO1 in human serum plasma tissue homogenates and cell lysates using a sensitive sandwich ELISA method It offers a broad detection range of 31 2 pg/mL to 2000 pg/mL with a high sensitivity of 7 8 pg/mL ensuring accurate quantification of low-abundance targets The assay requires a short incubation period of 1 to 5 hours and a modest sample volume of 50-100 L per test Detection is performed at 450 nm and the kit is designed for reliable performance in both research and clinical settings Its standardized protocol supports consistent results across diverse sample types making it suitable for studies focused on oxidative stress detoxification pathways or biomarker analysis related to GSTO1 expression