Learn More
Raybiotech Inc Human GSTP1 ELISA Kit
Supplier:  Raybiotech Inc ELHGSTP11
Encompass_Preferred
For Cell Culture Supernatants, Plasma, and Serum samples. Sensitivity: 0.41 ng/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA antibody pair detects human GSTP1. Other species not determined.
Catalog No. 50-194-9334