Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human GTF2IRD1 Full-length ORF (AAH18136, 1 a.a. - 959 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. p-7098132 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-946-812 25 μg
89-946-811 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-946-812 Supplier Abnova™ Supplier No. H00009569P01L
Only null left
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

The protein encoded by this gene contains five GTF2I-like repeats and each repeat possesses a potential helix-loop-helix (HLH) motif. It may have the ability to interact with other HLH-proteins and function as a transcription factor or as a positive transcriptional regulator under the control of Retinoblastoma protein. This gene is deleted in Williams-Beuren syndrome, a multisystem developmental disorder caused by deletion of multiple genes at 7q11.23. Alternative splicing of this gene generates at least 2 transcript variants. [provided by RefSeq]

Sequence: MALLGKRCDVPTNGCGPDRWNSAFTRKDEIITSLVSALDSMCSALSKLNAEVACVAVHDESAFVVSTEKGRMFLNARKELQSDFLRFCRGPPWKDPEAEHPKKVQRGEGGGRSLPRSSLEHGSDVYLLRKMVEEVFDVLYSEALGRASVVPLPYERLLREPGLLAVQGLPEGLAFRRPAEYDPKALMAILEHSHRIRFKLKRPLEDGGRDSKALVELNGVSLIPKGSRDCGLHGQAPKVPPQDLPPTATSSSMASFLYSTALPNHAIRELKQEAPSCPLAPSDLGLSRPMPEPKATGAQDFSDCCGQKPTGPGGPLIQNVHASKRILFSIVHDKSEKWDAFIKETEDINTLRECVQILFNSRYAEALGLDHMVPVPYRKIACDPEAVEIVGIPDKIPFKRPCTYGVPKLKRILEERHSIHFIIKRMFDERIFTGNKFTKDTTKLEPASPPEDTSAEVSRATVLDLAGNARSDKGSMSEDCGPGTSGELGGLRPIKIEPEDLDIIQVTVPDPSPTSEEMTDSMPGHLPSEDSGYGMEMLTDKGLSEDARPEERPVEDSHGDVIRPLRKQVELLFNTRYAKAIGISEPVKVPYSKFLMHPEELFVVGLPEGISLRRPNCFGIAKLRKILEASNSIQFVIKRPELLTEGVKEPIVDSQGTASSLGFSPPALPPERDSGDPLVDESLKRQGFQENYDARLSRIDIANTLREQVQDLFNKKYGEALGIKYPVQVPYKRIKSNPGSVIIEGLPPGIPFRKPCTFGSQNLERILAVADKIKFTVTRPFQGLIPKPDEDDANRLGEKVILREQVKELFNEKYGEALGLNRPVLVPYKLIRDSPDAVEVTGLPDDIPFRNPNTYDIHRLEKILKAREHVRMVIINQLQPFAEICNDAKVPAKDSSIPKRKRKRVSEGNSVSSSSSSSSSSSSNPDSVASANQISFVQWPMYMVDYAGLNVQLPGPLNY

Specifications

Accession Number AAH18136
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 9569
Molecular Weight (g/mol) 131.01kDa
Name GTF2IRD1 (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 25 μg
Immunogen MALLGKRCDVPTNGCGPDRWNSAFTRKDEIITSLVSALDSMCSALSKLNAEVACVAVHDESAFVVSTEKGRMFLNARKELQSDFLRFCRGPPWKDPEAEHPKKVQRGEGGGRSLPRSSLEHGSDVYLLRKMVEEVFDVLYSEALGRASVVPLPYERLLREPGLLAVQGLPEGLAFRRPAEYDPKALMAILEHSHRIRFKLKRPLEDGGRDSKALVELNGVSLIPKGSRDCGLHGQAPKVPPQDLPPTATSSSMASFLYSTALPNHAIRELKQEAPSCPLAPSDLGLSRPMPEPKATGAQDFSDCCGQKPTGPGGPLIQNVHASKRILFSIVHDKSEKWDAFIKETEDINTLRECVQILFNSRYAEALGLDHMVPVPYRKIACDPEAVEIVGIPDKIPFKRPCTYGVPKLKRILEERHSIHFIIKRMFDERIFTGNKFTKDTTKLEPASPPEDTSAEVSRATVLDLAGNARSDKGSMSEDCGPGTSGELGGLRPIKIEPEDLDIIQVTVPDPSPTSEEMTDSMPGHLPSEDSGYGMEMLTDKGLSEDARPEERPVEDSHGDVIRPLRKQVELLFNTRYAKAIGISEPVKVPYSKFLMHPEELFVVGLPEGISLRRPNCFGIAKLRKILEASNSIQFVIKRPELLTEGVKEPIVDSQGTASSLGFSPPALPPERDSGDPLVDESLKRQGFQENYDARLSRIDIANTLREQVQDLFNKKYGEALGIKYPVQVPYKRIKSNPGSVIIEGLPPGIPFRKPCTFGSQNLERILAVADKIKFTVTRPFQGLIPKPDEDDANRLGEKVILREQVKELFNEKYGEALGLNRPVLVPYKLIRDSPDAVEVTGLPDDIPFRNPNTYDIHRLEKILKAREHVRMVIINQLQPFAEICNDAKVPAKDSSIPKRKRKRVSEGNSVSSSSSSSSSSSSNPDSVASANQISFVQWPMYMVDYAGLNVQLPGPLNY
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias BEN/CREAM1/GTF3/MUSTRD1/RBAP2/WBS/WBSCR11/WBSCR12/hMusTRD1alpha1
Common Name GTF2IRD1
Gene Symbol GTF2IRD1
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.