Learn More
Biomatik Corporation Human Guanine Deaminase (GDA) ELISA Kit
Supplier:  Biomatik Corporation EKU04627
Encompass_Preferred
Human Guanine Deaminase (GDA) ELISA Kit for detecting GDA in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.129ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9Y2T3. Gene ID: 9615. Target Synonyms: CYPIN; Guanase; Guanine aminase; Guanine aminohydrolase; p51-nedasin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7589