Learn More
ACROBiosystems Human GUCY2C 25tests
Catalog No. 502534922
Encompass_Preferred
PE-Labeled Human GUCY2C Protein His Tag (GUC-HP2H9) is produced via conjugation of PE to Human GUCY2C Protein His Tag with a new generation site-specific technology under Star Staining labeling platform. Human GUCY2C Protein His Tag is expressed from human 293 cells (HEK293). It contains AA Ser 24 - Gln 430 (Accession # P25092-1).
Catalog No. 50-253-4922 Supplier ACROBiosystems Supplier No. GUCHP2H925TESTS
PE-Labeled Human GUCY2C Protein His Tag (GUC-HP2H9) is produced via conjugation of PE to Human GUCY2C Protein His Tag with a new generation site-specific technology under Star Staining labeling platform. Human GUCY2C Protein His Tag is expressed from human 293 cells (HEK293). It contains AA Ser 24 - Gln 430 (Accession # P25092-1).