Learn More
Sino Biological Anti-Glycophorin A Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 16018RP0120
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human GYPA / CD235a / Glycophorin A (rh GYPA / CD235a / Glycophorin A; Catalog#16018-H08H; P02724; Met1-Glu91). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-255-4073