Learn More
Sino Biological Anti-Granzyme B Antibody, Mouse Monoclonal
Supplier:  Sino Biological 10345MM33100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human Granzyme B (rh Granzyme B; Catalog#10345-H08H; NP_004122.1; Met1-Tyr247). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-255-5369