Learn More
BIOHIPPO Human GZMH Microsample
SDP
Catalog No. 5000315527
Encompass
Human GZMH Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of GZMH in Serum plasma tissue homogenates cell lysates and other biological fluids It provides a detection range of 31 25-2000 ng/mL with a sensitivity of 13 78 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human GZMH Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human GZMH Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human GZMH biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-15527 Supplier BIOHIPPO Supplier No. BHE15212499
Human GZMH Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of GZMH in Serum plasma tissue homogenates cell lysates and other biological fluids It provides a detection range of 31 25-2000 ng/mL with a sensitivity of 13 78 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human GZMH Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human GZMH Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human GZMH biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color