Learn More
BIOHIPPO Human GZMK Microsample
SDP
Catalog No. 5000314622
Encompass
Human GZMK Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of GZMK in serum plasma tissue homogenates cell lysates cell culture supernates and other biological fluids It provides a detection range of 0 32-20 ng/mL with a sensitivity of 0 112 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human GZMK Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human GZMK Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human GZMK biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-14622 Supplier BIOHIPPO Supplier No. BHE15212115
Human GZMK Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of GZMK in serum plasma tissue homogenates cell lysates cell culture supernates and other biological fluids It provides a detection range of 0 32-20 ng/mL with a sensitivity of 0 112 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human GZMK Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human GZMK Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human GZMK biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color