Learn More
BIOHIPPO Human GZMM Microsample
SDP
Catalog No. 5000315918
Encompass
Human GZMM Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of GZMM in Serum plasma tissue homogenates and other biological fluids It provides a detection range of 31 25-2000 ng/mL with a sensitivity of 13 52 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human GZMM Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human GZMM Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human GZMM biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-15918 Supplier BIOHIPPO Supplier No. BHE15212157
Human GZMM Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of GZMM in Serum plasma tissue homogenates and other biological fluids It provides a detection range of 31 25-2000 ng/mL with a sensitivity of 13 52 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human GZMM Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human GZMM Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human GZMM biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color