Learn More
Biomatik Corporation Human H2A Histone Family, Member X (H2AFX) CLIA Kit
Supplier:  Biomatik Corporation EKU11649
Encompass_Preferred
Human H2A Histone Family, Member X (H2AFX) CLIA Kit for detecting H2AFX in tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 27.4-20,000pg/ml. Sensitivity: 10.3pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: P16104. Gene ID: 3014. Target Synonyms: H2AX. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8106