Learn More
Biomatik Corporation Human H2A Histone Family, Member V (H2AFV) ELISA Kit
Supplier:  Biomatik Corporation EKU04632
Encompass_Preferred
Human H2A Histone Family, Member V (H2AFV) ELISA Kit for detecting H2AFV in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 1.56-100ng/ml. Sensitivity: 0.55ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q71UI9. Gene ID: 94239. Target Synonyms: H2AV. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6770