Learn More
Sino Biological Anti-MACROH2A1/H2AFY Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 104736T32100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Human H2AFY, and purified by antigen affinity chromatography.
Catalog No. 50-241-1595