Learn More
Sino Biological Anti-H3F3A Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 100815T32100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Human H3F3A, and purified by antigen affinity chromatography.
Catalog No. 50-240-9428