Learn More
BIOHIPPO Human HABP2 Microsample
SDP
Catalog No. 5000314308
Encompass
Human HABP2 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of HABP2 in Serum plasma and other biological fluids It provides a detection range of 6 25-400 ng/mL with a sensitivity of 2 52 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human HABP2 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human HABP2 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human HABP2 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-14308 Supplier BIOHIPPO Supplier No. BHE15211817
Human HABP2 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of HABP2 in Serum plasma and other biological fluids It provides a detection range of 6 25-400 ng/mL with a sensitivity of 2 52 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human HABP2 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human HABP2 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human HABP2 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color