Learn More
Raybiotech Inc Human HAI-1 ELISA Kit
Supplier:  Raybiotech Inc ELHHAI11
Encompass_Preferred
For cell culture supernatants, plasma, and serum samples. Sensitivity: 0.8 ng/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA antibody pair detects human HAI-1, no cross-reactivity with mouse HAI-1, human APP, GASP-1, GASP-2, HAI-2A, TFPI, TFPI-2 is observed.
Catalog No. 50-194-9338