Learn More
Sino Biological Human HAI-1 Matched ELISA Antibody Pair Set 5 Plates
Catalog No. 5000785182
Encompass_Preferred
The Human HAI-1/SPINT1 ELISA Pair Set is for the quantitative determination of Human HAI-1/SPINT1.The Sino Biological ELISA Pair Set is a solid phase sandwich ELISA (Enzyme-Linked Immunosorbent Assay). It utilizes a monoclonal antibody specific for Human HAI-1/SPINT1 coated on a 96-well plate. Standards and samples are added to the wells, and any Human HAI-1/SPINT1 present binds to the immobilized antibody. The wells are washed and a horseradish peroxidase conjugated rabbit anti-Human HAI-1/SPINT1 monoclonal antibody is then added, producing an antibody-antigen-antibody "sandwich". The wells are again washed and TMB substrate solution is loaded, which produces color in proportion to the amount of Human HAI-1/SPINT1 present in the sample. To end the enzyme reaction, the stop solution is added and absorbances of the microwell are read at 450 nm.
Catalog No. 50-007-85182 Supplier Sino Biological Supplier No. SEK117425
The Human HAI-1/SPINT1 ELISA Pair Set is for the quantitative determination of Human HAI-1/SPINT1.The Sino Biological ELISA Pair Set is a solid phase sandwich ELISA (Enzyme-Linked Immunosorbent Assay). It utilizes a monoclonal antibody specific for Human HAI-1/SPINT1 coated on a 96-well plate. Standards and samples are added to the wells, and any Human HAI-1/SPINT1 present binds to the immobilized antibody. The wells are washed and a horseradish peroxidase conjugated rabbit anti-Human HAI-1/SPINT1 monoclonal antibody is then added, producing an antibody-antigen-antibody "sandwich". The wells are again washed and TMB substrate solution is loaded, which produces color in proportion to the amount of Human HAI-1/SPINT1 present in the sample. To end the enzyme reaction, the stop solution is added and absorbances of the microwell are read at 450 nm.