Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human HAPLN2 Synthetic Peptide

Catalog No. PEP0617 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PEP0617 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PEP0617 Supplier Invitrogen™ Supplier No. PEP0617
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Synthetic Peptide

This peptide corresponds to 12 amino acids near the center of human Bral1. PEP-0617 can be used as a blocking peptide with polyclonal antibody PA5-20497.

BRAL1 is a member a superfamily consisting of several highly homologous hyaluronan and proteoglycan binding link proteins. BRAL1 is predominantly expressed in brain tissue and spinal cord. Like other members in the link-module superfamily, BRAL1 contains an immunoglobulin-like fold and two proteoglycan tandem repeats (PTRs). Its mRNA expression pattern is similar to other lectican proteoglycans, suggesting that BRAL1 may act to stabilize the binding between the extracellular matrix molecule hyaluronan and brevican. Immunostaining of mouse brain showed BRAL1 expression at P20 in the white matter of the developing cerebellum and in myelinated fiber tracts in the adult brain, suggesting that expression starts when axonal myelination occurs.
TRUSTED_SUSTAINABILITY

Specifications

Gene ID (Entrez) 60484
Species Human
Purification Method purified
Accession Number Q9GZV7
Common Name HAPLN2
Concentration 200 μg/mL
Content And Storage -20°C
Description Synthetic Peptide
Format Liquid
Formulation PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2
For Use With (Application) Blocking Assay, Control
Gene Alias 4930401E20Rik; Brain link protein 1; brain link protein-1; Bral1; HAPLN2; Hyaluronan and proteoglycan link protein 2; RP11-284F21.6
Gene Symbol HAPLN2
Lyophilized No
Preservative Sodium azide
Product Type Peptide
Quantity 50 μg
Regulatory Status RUO
Storage Buffer PBS
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.