Learn More
R&D Systems™ Human Haptoglobin Quantikine ELISA Kit



Description
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
| Assay Range | 3.13 - 200ng/mL |
| Assay Sensitivity | 0.529ng/mL |
| Conjugate | HRP |
| Sample Type | Plasma (EDTA, Heparin), Saliva, Serum, Serum-free Cell Culture Supernatant, Urine |
| Specificity | Natural Human Haptoglobin |
| Sufficient For | 1 x 96-well Plate |
| For Use With (Application) | ELISA |
| Gene ID (Entrez) | 3240 |
| Sample Volume | Serum-free Cell Culture Supernates (20uL),Serum (10uL),EDTA Plasma (10uL),Heparin Plasma (10uL),Saliva (10uL),Urine (20uL) |
| Storage Requirements | Store the unopened product at 2° to 8°C. Do not use past expiration date. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.