Learn More
Biomatik Corporation Human Haptoglobin Related Protein (HPR) CLIA Kit
Supplier:  Biomatik Corporation EKU09928
Encompass_Preferred
Human Haptoglobin Related Protein (HPR) CLIA Kit for detecting HPR in serum, plasma, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 68.6-50,000pg/ml. Sensitivity: 25.4pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: P00739. Gene ID: 3250. Target Synonyms: HP. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7647