Learn More
Sino Biological Recombinant Human HARS/HisRS Protein (His Tag)
Supplier:  Sino Biological 10172H07B100
Encompass_Preferred
A DNA sequence encoding the human HARS (NP_002100.2) (Ala2-Cys509) was expressed with a polyhistidine tag at the N-terminus.
Catalog No. 50-255-5444